Principaux documents
TMO2401-0500
Nalgene® economy wash bottle
low-density polyethylene, volume 500 mL, unlabelled, case of 24 ea
Synonyme(s) :
LDPE flush bottle, LDPE wash bottle
24 EACH
242,00 $
242,00 $
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Changer de vue
24 EACH
242,00 $
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.43
242,00 $
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Matériaux
low-density polyethylene
Conditionnement
case of 24 ea
Fabricant/nom de marque
Nalgene 2401-0500
Capacité
500 mL
Volume
500 mL
Description générale
Prevent separation when squeezing Thermo Scientific Nalgene LDPE Economy Wash Bottles, featuring a tight-fitting draw tube and closure. These low density polyethylene bottles are designed to dispense through the top with a closure and stem molded in one piece for uniform leakproof† service. Cut back tip to increase flow.
Informations légales
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries