Principaux documents
S5939
Sigma-Aldrich
Sun cap closures
Biofilter, O.D. 18 mm
About This Item
Code UNSPSC :
41100000
Description
disposable
Caractéristiques
autoclavable
D.E.
18 mm
Application(s)
agriculture
Description générale
8 mm 0.04 μm polypropylene filter disc with heatproof adhesive. Comes with 20 closures per sheet and 25 total sheets.