S5894
Sigma lab markers
Fine tip, black
About This Item
Code UNSPSC :
41100000
Produits recommandés
Couleur
black
Description générale
Writes on cold, wet surfaces. Quick-drying. Alcohol based permanent marker removed by alcohol.