Documents
PURNR
Sigma-Aldrich
Pur-A-Lyzer Floating Racks for Dialysis
Mini
Synonyme(s) :
Dialyzers, High through put dialysis tubing
About This Item
Code UNSPSC :
41122804
Nomenclature NACRES :
NB.22
Conditionnement
pack of 1 ea
Description générale
Floating rack can process up to 63 Mini Pur-A-Lyzer Dialysis Tubes in a single beaker.
Informations légales
Pur-A-Lyzer is a trademark of Sigma-Aldrich Co. LLC