Principaux documents
MTCB8300
MTC™ Bio Collapsible Bio Bucket
laboratory ice bucket
Synonyme(s) :
sample cooling, sample storage
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.15
Matériaux
polystyrene
Conditionnement
pack of 1 ea
Fabricant/nom de marque
MTC Bio B8300
P × H
10 in. × 7.5 in.
Description générale
Bio Bucket collapsible laboratory ice bucket, 5 Liter, 1/each
A collapsible container that conserves storage space. BioBucket can be used as an ice bucket to keep samples cool over long periods of time or as a portable liquid container. Attached handle for transportation and the molded base grip allows for easy pouring and cleaning. Ideal for field use as an environmental sample collection and transport receptacle.
Caractéristiques et avantages
  • Collapsible body saves valuable storage space
  • Useable with crushed ice
  • Inner-lid access port for enhanced temperature retention
  • Attached handle for easy transportation
  • Molded grip on base for pouring
Informations légales
BioBucket is a trademark of MTC Bio
MTC is a trademark of MTC Bio