Principaux documents
HS15989A
Premium Slide box
25 Place, cork lining, blue
Synonyme(s) :
Schiebekasten, slide box microscope, slide storage, slide storage box
About This Item
Code UNSPSC :
41123302
Nomenclature NACRES :
MA.01
Le tarif et la disponibilité ne sont pas disponibles actuellement.
Matériaux
blue
nickel closure
Caractéristiques
Holds 25 x slides 76x26 mm
Fabricant/nom de marque
Heathrow Scientific HS15989A
L × l × H
208 × 175 × 34 mm
Description générale
Slide boxes are perfect for storage or transportation of standard size slides ( 1 x 3in.).
Caractéristiques et avantages
  • molded, rigid boxes securely hold slides in grooved slots
  • durable ABS plastic boxes are available wth either cork or foam lining
  • both the lining and the slots are numbered to correspond to the slide inventory sheet
  • closes securely with a rust-resistant, nickel-plated clasp and pin hinges