HS15994M
Premium slide box
100 place, foam, blue plastic
Synonyme(s) :
Schiebekasten, slide box microscope, slide storage, slide storage box
35,80 $
About This Item
35,80 $
Produits recommandés
Matériaux
blue plastic
nickel closure
Caractéristiques
Holds 100 x slides 76x26 mm
Fabricant/nom de marque
Heathrow Scientific HS15994M
L × l × H
208 × 175 × 34 mm
Catégories apparentées
Caractéristiques et avantages
- Molded, rigid boxes securely hold slides in grooved slots
- Durable ABS plastic boxes are available with cork lining only
- Both the lining and the slots are numbered to correspond to the slide inventory sheet
- Feature thumb-latch lock mechanism