HS120249
Wide mouth wash bottle
capacity 500 mL, blue PE-LD bottle
50,40 $
About This Item
50,40 $
Produits recommandés
Matériaux
blue PE-LD bottle
polypropylene cap
Caractéristiques
label no label
vented
with cap
Conditionnement
pack of 6 ea
Fabricant/nom de marque
Heathrow Scientific HS120249
Capacité
500 mL
Catégories apparentées
Description générale
The closure and stem are molded in one piece to make them leak-proof. The angled neck and tip provide a uniform stream of liquid and the tip can be cut back if required to increase the flow.
- Wide bottle mouth makes filling easier
- Color-coding for user, application or location identification
- Bottle is made from LDPE, with a PP closure
- 500 mL bottle size,
- Supplied in packs of 6