Principaux documents
GE80-1129-37
GelBond PAGfilm, 203 X 260 mm, Pkg/50
Cytiva 80-1129-37, pack of 50 pieces
1 PKG
903,00 $
903,00 $
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Changer de vue
1 PKG
903,00 $
About This Item
Code UNSPSC :
41122410
Nomenclature NACRES :
NB.22
903,00 $
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Conditionnement
pack of 50 pieces
Fabricant/nom de marque
Cytiva 80-1129-37
Disponibilité
not available in Japan
Température de stockage
room temp
Catégories apparentées
Description générale
GelBond PAGfilm is designed as a support for polyacrylamide gels. One side of the film has a specially treated surface which will react with the acrylamide monomer solution during polymerization to form covalent linkages between the gel and the plastic support.
Stockage et stabilité
Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.
Remarque sur l'analyse
To view the Certificate of Analysis for this product, please visit www.cytiva.com.