Principaux documents
GE29-0230-51
Frac30 fraction collector
round
About This Item
Code UNSPSC :
41105300
Nomenclature NACRES :
NA.55
Le produit GE29-0230-51 n'est pas actuellement commercialisé dans votre pays Contacter notre Service technique
Matériaux
round
Fabricant/nom de marque
Cytiva 29-0230-51
Catégories apparentées
Description générale
Frac30 fraction collector is controlled via the ÄKTA start touch screen instrument display or using UNICORN start software on a computer and 30 allows you to collect up to 30 fractions and supports four tube sizes, ranging from 1.5 mL to 15 mL.
Fractions can be collected in volumes ranging from 0.5 mL to 15 mL. The entire fraction collector rack can be transferred as required.
available in Europe only