Principaux documents
GE28-9068-42
Hyperfilm MP
W × L 5 in. × 7 in., pack of 50 ea
1 PKG
469,00 $
469,00 $
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Changer de vue
1 PKG
469,00 $
About This Item
Code UNSPSC :
41105300
Nomenclature NACRES :
NB.22
469,00 $
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Conditionnement
pack of 50 ea
Fabricant/nom de marque
Cytiva 28906842
Larg. × L
5 in. × 7 in.
Description générale
Amersham Hyperfilm MP is an excellent multipurpose film constructed from a clear base coated on both sides with a photographic emulsion that is protected by an anti-scratch layer. The anti-scratch coating reduces the risk of visible artifacts (e.g., plastic wrap marks) and the clear colorless base results in easy reading of images, without compromising on sensitivity.
Caractéristiques et avantages
  • High sensitivity for low concentration targets.
  • Intended for radiometric or chemiluminescent applications.
  • Clear background for excellent contrast and improved band visibility.
  • Publication-quality images.
  • High resolution at an economical price.
  • Easy to use.
Stockage et stabilité
Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.
Informations légales
Hyperfilm is a trademark of Cytiva