Principaux documents
EP0030128788
Eppendorf® Twin tec PCR plates, 96 wells
yellow plate, conical bottom colorless wells, skirted, pkg of 300 ea
About This Item
Code UNSPSC :
41106300
Nomenclature NACRES :
NB.15
Matériaux:
conical bottom colorless wells
yellow plate
Taille:
96 wells
Volume maximum des puits:
150 μm
Caractéristiques:
skirt
Le produit EP0030128788 n'est pas actuellement commercialisé dans votre pays Contacter notre Service technique
Matériaux
conical bottom colorless wells
yellow plate
Caractéristiques
skirt
Conditionnement
pkg of 300 ea
Fabricant/nom de marque
Eppendorf® 0030128788
Technique(s)
PCR: suitable
Taille
96 wells
Volume maximum des puits
150 μm
Description générale
  • Eppendorf® Twin tec PCR plates, 96 wells are single-use polycarbonate/polypropylene plates allowing trained specialists to perform PCR and cycle sequencing experiments.
  • They are used in training, routine and research laboratories in the bioscientific, industrial and chemical fields.
  • Evaporation is prevented with the aid of a variety of sealing systems.
Choose Eppendorf twin.tec PCR plates for confidence in every PCR assay. twin.tec plates offer purity, rigidity, color, and other simple, smart features can make your daily life a lot easier. The rigid polycarbonate frame works well in automation and high-throughput processes, and the different colors mean you can quickly organize the different steps of your workflow.
Informations légales
Eppendorf is a registered trademark of Eppendorf AG