Principaux documents
EP0030127692
Eppendorf® Serological Pipets, Sterile Grade
yellow, capacity 1 mL, pack of 800 (4 bags × 200 pcs.)
800 EA
323,00 $
323,00 $
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Changer de vue
800 EA
323,00 $
About This Item
Code UNSPSC :
41121500
Nomenclature NACRES :
NB.04
323,00 $
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Matériaux
yellow
Stérilité
sterile
Caractéristiques
DNA free
graduations
plugged
Conditionnement
pack of 800 (4 bags × 200 pcs.)
Fabricant/nom de marque
Eppendorf® 0030127692 (Global)
Capacité
1 mL
Description générale
  • Eppendorf® Serological Pipets are manufactured from ultra-pure virgin polystyrene for optimal clarity.
  • Easy-to-read graduations for accurate volume determination.
  • Pipets are color-coded based on sizes for easy identification.
  • Sterility assurance level of 10-6.
  • Compatible with all mechanical and electronic pipette controllers - including Pipet Helper® or Easypet® 3.
  • Free of detectable pyrogens, DNA, RNase and DNase and certified non-cytotoxic.
Application
  • Transfer of liquids
  • Mixing suspensions
  • Combining reagents and chemical solutions
  • Transferring cells for empirical analysis or expansion
  • Layering reagents for creating higher density gradients
Informations légales
Easypet is a registered trademark of Eppendorf AG
Eppendorf is a registered trademark of Eppendorf AG
Pipet Helper is a registered trademark of Eppendorf AG