Key Documents
OMK (637-69)
90110510, owl monkey kidney, Not specified
About This Item
Code UNSPSC :
41106514
product name
OMK (637-69), from owl monkey kidney, 90110510, Convention on the International Trade in Endangered Species of Wild Fauna and Flora (CITES)
Source biologique
owl monkey kidney
Mode de croissance
Adherent
Caryotype
Not specified
Morphologie
Not specified
Récepteurs
Not specified
Origine de la lignée cellulaire
Owl Monkey Kidney
Description de la lignée cellulaire
Derived from a normal female adult owl monkey. Sub-cultivation on over 100 occasions produced no evidence of latent infection. Used for propagation of non-human primate viruses, e.g., Herpesvirus aotus, Herpesvirus saimari and Herpesvirus ateles.
Milieu de culture
EMEM (EBSS) (M2279) + 2 mM L-Glutamine (G7513) + 1% NEAA (M7145) + 10% FBS / FCS (F2442).
Procédure de repiquage
Split sub-confluent cultures (70-80%) 1:4 to 1:8, i.e., seeding at 1-3x10,000 cells/cm2 using 0.25% trypsin; 5% CO2; 37 °C.
Autres remarques
Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.