Principaux documents
BMSW100520
Sigma-Aldrich
Accuris M1 Grade Calibration Weight
size 20 g, stainless steel
About This Item
Code UNSPSC :
41100000
Nomenclature NACRES :
NB.71
Matériaux
stainless steel
Conditionnement
pack of 1 ea
Fabricant/nom de marque
Benchmark Scientific W1005-20
Taille
20 g
Description générale
  • F1 Grade tolerance per OIML (International Organization of Legal Metrology)
  • Works with all Accuris Lab balances
  • Non-Magnetic Stainless Steel
  • Includes a protective plastic case
Informations légales
Accuris is a trademark of Benchmark Scientific, Inc.