Principaux documents
BAF198070000
Bel-Art® Centrifuge Tubes, Rimless
tube capacity × L 45 mL × 104 mm, polypropylene, pack of 12 tubes
Synonyme(s) :
round bottom tube
About This Item
Code UNSPSC :
41121703
Nomenclature NACRES :
NB.22
Matériaux
polypropylene
round bottom
Caractéristiques
autoclavable
Conditionnement
pack of 12 tubes
Fabricant/nom de marque
Bel-Art F19807-0000
Tube, capacité × L
45 mL × 104 mm
Description générale
Rimless polypropylene centrifuge tubes for use in centrifuges up to 50,000 xg without refrigeration. Autoclavable at 121ºC (250ºF).
Informations légales
Bel-Art is a registered trademark of SP Scienceware, a division of SP