Principaux documents
Z801240
Hellma® aluminum spacer
38mmx12.5mmx8 mm, to fit cells with 2 mm light path into 10 mm cell holder
About This Item
Code UNSPSC :
12352200
Nomenclature NACRES :
SB.32
Fabricant/nom de marque
Hellma 13.102
Technique(s)
UV/Vis spectroscopy: suitable
photometry: suitable
Informations légales
Hellma is a registered trademark of Hellma GmbH