Principaux documents
Z743167
Julabo® bath covers
flat cover to fit CORIO CD-B39
1 EA
649,00 $
649,00 $
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Changer de vue
1 EA
649,00 $
About This Item
Code UNSPSC :
41103706
Nomenclature NACRES :
NB.77
649,00 $
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Conditionnement
pack of 1 ea
Fabricant/nom de marque
Julabo 9970293
Description générale
Flat stainless steel bath cover reduces the evaporation of the bath fluid and saves energy. The rising steam is led back into the bath. This means that the bath fluid remains in the bath and refill cycles are extended. An additional benefit is that the cover prevents contamination of the samples in the bath.
Informations légales
Julabo is a registered trademark of Julabo USA, Inc.