Principaux documents
Z742353
2MAG Stirring bar remover XL
with integrated corrosion-resistant high-energy magnet
Synonyme(s) :
magnetic extraction tool
1 EA
1 540,00 $
1 540,00 $
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Changer de vue
1 EA
1 540,00 $
About This Item
Code UNSPSC :
41103806
Nomenclature NACRES :
NB.72
1 540,00 $
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Fabricant/nom de marque
2mag 45100
Description générale
Extremely strong stirring bar remover with an integrated corrosion-resistant high energy magnet, ideal for reliable inserting and removing of strong and heavy magnetic stirring bars with a weight up to 1,000 g. Allows for simple handling due to a special developed stripping mechanism, constructed from stainless steel. Solid and easy to clean with high chemical resistance. Length of 100 cm for use in deep reaction vessels.