Principaux documents
Z742265
DURAN® GL 56 Bottle Tag
blue silicone
Synonyme(s) :
292435659
20 EA
141,00 $
141,00 $
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Changer de vue
20 EA
141,00 $
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.17
141,00 $
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Matériaux
blue silicone
Conditionnement
pack of 20 ea
Fabricant/nom de marque
DURAN® 292435659
Description générale
Bottle tags can be used on their own for color identification or to secure the protective light sheild around the bottle. The GL 56 bottle tags are available in four colors (orange, yellow, blue and purple).
Caractéristiques et avantages
  • Manufactured from silicone rubber
  • The Bottle Tags can be used for colour identification or to secure the protective Light Shield around the bottle
  • The elastic Bottle Tags can be easily and securely attached around the neck, even with the screw cap in place
  • Feature a graspable tab that makes them easy to remove with bare or gloved hands
  • Chemically robust, heat resistant and reusable
Informations légales
DURAN is a registered trademark of DWK Life Sciences