Principaux documents
Z683280
J.Young air inlet valve, 5mm bore
pkg of 1 ea
About This Item
Code UNSPSC :
40141600
Nomenclature NACRES :
NB.79
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
J. Young
Description générale
The J. Young air inlet valve helps in controlling the atmosphere when using air-sensitive materials.