Z675598
Polyethylene carboy with spigot
size 8 L (2 gal)
279,00 $
About This Item
279,00 $
Produits recommandés
Matériaux
low-density polyethylene carbonate (Bottle and spigot)
Conditionnement
pack of 1 ea
Fabricant/nom de marque
Bel-Art F11847-0020
Taille
8 L (2 gal)
Catégories apparentées
Description générale
- Spigot opens fully in one half-turn
- Molded of polyethylene, which resists corrosive acids and alkalis
- Extra heavy walls, yet lighter than glass