Principaux documents
Z567590
Sigma-Aldrich
Aldrich® Benchclamp 2-prong
dual adjustment grippy knobs, size S
Synonyme(s) :
bench clamps
About This Item
Code UNSPSC :
41122413
Nomenclature NACRES :
NB.35
Le tarif et la disponibilité ne sont pas disponibles actuellement.
Taille
S
Capacité de la mâchoire
0-40 mm
Largeur de la fixation
6 mm
Description générale
Aldrich benchclamp clamps have a nickel-plated finish and are corrosion resistant. The clamps come with slip-on fiberglass finger covers. The Grippy Knob design has a round nut which is easy to turn making tightening the clamps around glassware fast and convenient.
Informations légales
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Benchclamp is a trademark of Sigma-Aldrich Co. LLC