Z177288
Stainless steel clamp
Overall L 8 1/2 in.
Conditionnement
pack of 1 ea
Fabricant/nom de marque
Bel-Art H18354-0000
L totale
8 1/2 in.
Description générale
Tri-grip, 0 to 1 in. opening range. Has spring-action opening with flat wing-head screw for clamping. Jaws covered with rubber tubing.