Principaux documents
Z531375
Sigma-Aldrich
Aldrich® Graham condenser
Overall H 200 mm, joint: ST/NS 14/20
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.43
Joint
joint: ST/NS14/20
Disponibilité
not available in EU
H totale
200 mm
Description générale
The Graham condenser has a large cooling surface area by allowing coolant to circulate around the inner coil. Condensers have a standard taper outer joint and a lower inner drip-tip joint.
Informations légales
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC