Principaux documents
Z400165
Sigma-Aldrich
Fine test sieves
SS frame, pore size 63 μm
1 EA
205,00 $
205,00 $
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Changer de vue
1 EA
205,00 $
About This Item
Code UNSPSC :
41105003
Nomenclature NACRES :
NB.35
205,00 $
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Matériaux
stainless steel
Stérilité
non-sterile
Conditionnement
pack of 1 ea
Fabricant/nom de marque
ATM 230SS8F
Taille des particules
230 mesh
Dimension de pores
63 μm
Description générale
  • Full 2 in. height and 8 in. diameter test sieves
  • Meet ASTM specification E-11
  • SS wire cloth measurement traceable to NIST
  • Each sieve includes a certificate of compliance to all applicable ASTM, ISO, and ANSI specifications
These metal framed testing sieves are manufactured with a lead/tin solder on the underside which joins the wire cloth to the sieve skirt. The top seam is secured with a UV cured adhesive.
Contenu apparenté
Particle size conversion table from our Lab Basics Technical Library