Principaux documents
Z314366
FORTUNA® Optima glass syringes with interchangeable components
capacity 20 mL, glass Luer tip style
About This Item
Code UNSPSC :
42142600
eCl@ss :
32070500
Nomenclature NACRES :
NB.03
Matériaux
borosilicate glass
Caractéristiques
autoclavable
graduated 1 mL
Fabricant/nom de marque
Fortuna Graf
Capacité
20 mL
Description générale
  • Interchangeable barrels and plungers
  • Flanges flat on two sides to prevent syringe rolling
  • Colored ring on plunger ends for easy volume measurement
  • Conformity certified by H mark
Glass Luer
- standard Luer hub
Luer lock - metal Luer lock tip on glass syringe, easy twist locks Luer needle onto syringe
Informations légales
FORTUNA is a registered trademark of Poulten & Graf Ltd.