There is an inside height of 89mm, and inner diameter of 69.5mm.
Z137065
Jacketed beaker
275,00 $
About This Item
275,00 $
Produits recommandés
Matériaux
borosilicate glass
Caractéristiques
graduations: no
Volume
250 mL
-
Hi! Could you please detail the inside diameter of the beaker?