Principaux documents
Z303909
Quickfit® flat flange lids for reaction flasks
center joint: ST/NS 19/26, parallel socket joint: 19/26, 5° side socket joint: 24/29, 10° side socket joint: 19/26
Synonyme(s) :
MAF2/32
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.43
Le produit Z303909 n'est pas actuellement commercialisé dans votre pays Contacter notre Service technique
Matériaux
borosilicate glass
Étiquette/Marqueur
A
Joint
10° side socket joint: 19/26
15° side socket joint
5° side socket joint: 24/29
center joint: ST/NS19/26
parallel socket joint: 19/26
Stérilité
non-sterile
Conditionnement
pack of 1
Fabricant/nom de marque
Quickfit MAF 2/32
Disponibilité
not available in USA and Canada
Description générale
Single and multi-socket lids for reaction flasks for simple extraction, reflux or for the introduction of a variety of stirrers, dropping funnels, temperature probes etc.
Use clip to secure lid to flasks, purchase separately.
Informations légales
Quickfit is a registered trademark of SciLabware Limited