Documents
Z301337
Quickfit® plain bends
joint: ST/NS 29/32
Synonyme(s) :
RA9/44
About This Item
Code UNSPSC :
41121815
Nomenclature NACRES :
NB.35
Joint
joint: ST/NS29/32
Fabricant/nom de marque
Quickfit RA 9/44
Disponibilité
not available in USA and Canada
not available in USA and Canada
Description générale
Socket to cone. Generally used as distilling adapters.
Informations légales
Quickfit is a registered trademark of SciLabware Limited