Z300896
Quickfit® Reduction Adapters
top female joint: ST/NS 14/23, bottom male joint: ST/NS 29/32
Synonyme(s) :
DA14
About This Item
Produits recommandés
Joint
female joint: ST/NS top14/23
male joint: ST/NS bottom29/32
Fabricant/nom de marque
Quickfit DA 14
Disponibilité
not available in USA and Canada
Catégories apparentées
Description générale
These adapters connect items with a smaller cone to a piece of glassware with a larger size socket.
Informations légales
Quickfit is a registered trademark of SciLabware Limited