Principaux documents
Z284181
Spinvane® magnetic stirring bar
for centrifuge tubes
Synonyme(s) :
Stir Bar, Stirbar, Stirring Bar
About This Item
Code UNSPSC :
41103806
Matériaux
PTFE-coated (Half round tapered)
PTFE-coated
Conditionnement
pack of 5 ea
Fabricant/nom de marque
Bel-Art F37137-0000
Barre, Larg. × Haut. × Diam.
9.7/6.1 mm × 23.0 mm × 9.9 mm
Description générale
  • Spinvane® magnetic stirring bars are ideal for small conical-shaped tubes and micro vials.
  • The triangle-shaped polytetrafluoroethylene (PTFE) coated vane features a cross-mounted molded-in Alnico magnet for effective stirring.
  • Autoclavable.
Bars designed for narrow stirring applications.
Informations légales
Spinvane is a registered trademark of SP Scienceware, a division of SP