The caps for this item do have a ring in them for tight seals. The supplier has tested the bottles and deemed them leak proof.
Z261106
Nalgene® PassPort™ IP2 bottles
Narrow-mouth, capacity 500 mL
Synonyme(s) :
nalgene bottle, nalgene bottles, nalgene hdpe
135,00 $
About This Item
135,00 $
Produits recommandés
Matériaux
high-density polyethylene
narrow-mouth bottle
polypropylene cap
round bottle
translucent bottle
Joint
Joints threaded neck (28-415)
Caractéristiques
with cap
Conditionnement
pack of 12 (bottles)
Fabricant/nom de marque
Nalgene® 2099-0016
Capacité
500 mL
Diam. × H
73 mm × 171 mm
-
Hello, regarding Nalgene PassPort IP2 bottles, do the caps have a seal ring at the top providing a tight seal?