Principaux documents
Z225320
Sigma-Aldrich
Aldrich® SAFE-PURGE valve
manual
About This Item
Code UNSPSC :
40141600
Nomenclature NACRES :
NB.79
Le tarif et la disponibilité ne sont pas disponibles actuellement.
Paramètres
<5 psi pressure
Description générale
Used with purge gas pressure under 5 psi.
The Aldrich SAFE-PURGE valves provide for safe, high-performance operation. Both the manual and automatic spring-loaded styles feature a durable PTFE metering valve.
  • Quick and efficient purging of reaction vessel which conserves expensive purge gases
  • Safely vents toxic gases from built-in oil bubbler
Informations légales
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
SAFE-PURGE is a trademark of Sigma-Aldrich Co. LLC