Principaux documents
Z188824
Supelco
Black plastic caps
Solid top, white rubber liner, thread for 20-400, pkg of 500 ea
About This Item
Code UNSPSC :
41122001
Nomenclature NACRES :
NB.11
Matériaux
black phenolic cap
white rubber liner
Caractéristiques
closure type black phenolic solid top cap
Conditionnement
pkg of 500 ea
Raccords
thread for 20-400
Description générale
These autoclavable caps are designed to ensure the ultimate in product safety.