Z158321
Ace Reducing Adapter
female joint: ST/NS 24/40, male joint: SJ 35/25
About This Item
Code UNSPSC :
41121815
Nomenclature NACRES :
NB.35
Produits recommandés
Joint
female joint: ST/NS24/40
male joint: SJ35/25
Fabricant/nom de marque
Ace Glass 502521