Principaux documents
Z126748
Stainless steel 304 syringe needle
gauge 14, L 24 in.
1 EA
107,00 $
107,00 $
En stockDétails
Changer de vue
1 EA
107,00 $
About This Item
Code UNSPSC :
41122002
Nomenclature NACRES :
NB.03
107,00 $
En stockDétails
Description
Standard Hub with 304 Stainless Steel Tubing
Conditionnement
pack of 1 ea
Fabricant/nom de marque
Cadence Science
Longueur
24 in.
Calibre
14
Description générale
304 SS needle with chrome-plated brass Luer hub. Needle is designed with a slightly curved, deflecting tip to prevent coring during needle insertion through the septum.
Standard Hub with 304 Stainless Steel Tubing