Principaux documents
WHA10442100
Whatman® glass vacuum filtration device
GV 050/1, sieve filter, rubber stopper connection, 1/pk
Synonyme(s) :
Z698334, glass vacuum filtration unit, vacuum filtration system
1 PKG
968,00 $
968,00 $
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Changer de vue
1 PKG
968,00 $
About This Item
Code UNSPSC :
41104900
Nomenclature NACRES :
NB.24
968,00 $
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Matériaux
borosilicate glass funnel(s) (funnel)
borosilicate glass housing
stainless steel clamp(s) (clamp)
Caractéristiques
glass vacuum filter holder, 47/50mm support screen
Conditionnement
pack of 1
Fabricant/nom de marque
Whatman 10442100
Whatman Article No. 28415561 (US reference)
Capacité de l'entonnoir
250 mL
Adéquation
suitable for (filtration and biological analysis; the 304 stainless steel support screen is suitable for use with proteinaceous solutions)
Description générale
Whatman® Vacuum Type Glass Holders
Produced from borosilicate glass and available with a choice of support screen. Suitable for aqueous and organic solvent filtration. The funnel seal ensures that the sample does not bypass the membrane and that particulates are retained on the surface of the membrane.
The sintered glass support is recommended for filtration and biological analysis. The 304 stainless steel support screen is suitable for use with proteinaceous solutions.
Caractéristiques et avantages
  • Chemically resistant to most aqueous and organic solutions.
  • Resistant to acids and caustic solutions.
  • Autoclavable and can be sterilized in dry heat to 18NA°C.
  • Can be used up to 200°C.
Autres remarques
Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.
Informations légales
Whatman is a registered trademark of Cytiva