Principaux documents
W508101
Sigma-Aldrich
1-Methyl-1,4-cyclohexadiene
≥96%, stabilized
About This Item
Formule empirique (notation de Hill):
C7H10
Numéro CAS:
Poids moléculaire :
94.15
Numéro CE :
Numéro MDL:
Code UNSPSC :
12164502
ID de substance PubChem :
Nomenclature NACRES :
NA.21
Propriétés organoleptiques:
faint
Source biologique:
synthetic
Allergène alimentaire:
no known allergens
Plus de détails sur ce produit W508101 Contacter notre Service technique
Source biologique
synthetic
Niveau de qualité
Essai
≥96%
Contient
BHT as stabilizer
Indice de réfraction
n20/D 1.471 (lit.)
pb
114-115 °C (lit.)
Densité
0.838 g/mL at 25 °C (lit.)
Application(s)
flavors and fragrances
Documentation
see Safety & Documentation for available documents
Allergène alimentaire
no known allergens
Propriétés organoleptiques
faint
Chaîne SMILES 
CC1=CCC=CC1
InChI
1S/C7H10/c1-7-5-3-2-4-6-7/h2-3,6H,4-5H2,1H3
Clé InChI
QDXQAOGNBCOEQX-UHFFFAOYSA-N
Description générale
1-Methyl-1,4-cyclohexadiene is reported to be produced by some Nodulosporium species along with other volatile organic compounds (VOCs).[1][2]
Pictogrammes
FlameExclamation mark
Mention d'avertissement
Danger
Mentions de danger
Classification des risques
Eye Irrit. 2 - Flam. Liq. 2
Code de la classe de stockage
3 - Flammable liquids
Classe de danger pour l'eau (WGK)
WGK 3
Point d'éclair (°F)
51.8 °F - closed cup
Point d'éclair (°C)
11 °C - closed cup
János P Mészáros et al.
Dalton transactions (Cambridge, England : 2003), 49(23), 7977-7992 (2020-06-06)
Herein the design and synthesis of a new 8-hydroxyquinoline derivative, (S)-5-chloro-7-((proline-1-yl)methyl)8-hydroxyquinoline (HQCl-Pro), with good water solubility and multidrug resistance reversal activity are reported. In this work the proton dissociation processes of HQCl-Pro and its complex formation with [Rh(η5-C5Me5)(H2O)3]2+, [Ru(η6-p-cymene)(H2O)3]2+ and
An endophytic Nodulisporium sp. producing volatile organic compounds having bioactivity and fuel potential.
Mends MT, et al.
Journal of Petroleum & Environmental Biotechnology, 3(3) (2012)
Hypoxylon sp., an endophyte of Persea indica, producing 1, 8-cineole and other bioactive volatiles with fuel potential.
Tomsheck AR, et al.
Microbial Ecology, 60(4), 903-914 (2010)