Principaux documents
TEM-7596408
Sigma-Aldrich
OMNI Probe Copper 3-Post Lift-Out Grid
pack of 100 (per vial)
Synonyme(s) :
Lift-Out OMNI GRIDS, 3-Post, Shallow Downset, Copper
About This Item
Code UNSPSC :
41111761
Nomenclature NACRES :
MA.24
Le produit TEM-7596408 n'est pas actuellement commercialisé dans votre pays Contacter notre Service technique
Conditionnement
pack of 100 (per vial)
Description générale
3 post copper lift-out grids specifically designed for in-situ lift-out. These grids include multiple indexed mounting locations with both vertical bar and "V" shaped attachment surfaces. These grids have a shallower downset and slightly wider center post.
Conditionnement
Packaged in glass vials.