Principaux documents
SYNV163024
Synthware vacuum trap with serrated hose connections
joint: ST/NS 24/40, body O.D. 30 mm, L 200 mm , below joint
Synonyme(s) :
Synthware V163024
1 EA
143,00 $
143,00 $
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Changer de vue
1 EA
143,00 $
About This Item
Code UNSPSC :
40161501
Nomenclature NACRES :
NB.51
143,00 $
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Joint
joint: ST/NS24/40
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
Synthware V163024
Longueur
200 mm , below joint
D.E. du corps
30 mm
Catégories apparentées
Description générale
With joints for easy removal of condensate. Serrated hose connections for side tube have an O.D. of 13 mm at the smallest serration and 16 mm at the largest serration. Center tube hose connection is always 10mm for easy connection to manifold bank tubes which are also with 10 mm hose connection.
Informations légales
Synthware is a trademark of Kemtech America Inc.