Principaux documents
SYNS181635
Synthware straight bore PTFE stopcock plug
bore size 6 mm, Plug Size: 16/35
Synonyme(s) :
Synthware S181635
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.51
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
Synthware S181635
Taille de l'alésage
6 mm
Description générale
This replacement kit is complete with one PTFE plug (1:5 taper), one PTFE washer, one O-ring.
Informations légales
Synthware is a trademark of Kemtech America Inc.