Documents
SYNF434950
Synthware three neck round bottom flask with angled side necks
50 mL, center joint: ST/NS 24/40, side joint: ST/NS 19/22
Synonyme(s) :
Synthware F434950, 3-neck round flask
About This Item
Code UNSPSC :
41121804
Nomenclature NACRES :
NB.51
Matériaux
borosilicate glass flask
round bottom
three-neck (angled side necks)
Joint
center joint: ST/NS24/40
side joint: ST/NS19/22
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
Synthware F434950
Capacité
50 mL
50 mL
Description générale
Round bottom with joints. Heavy wall design. All sizes are hand-blown from tubing to ensure uniform wall thickness.
Informations légales
Synthware is a trademark of Kemtech America Inc.