Principaux documents
SYNF352440M
Synthware fritted filter tube with glass stopcocks
joint: ST/NS 24/40, disc O.D. 36 mm, porosity: medium
Synonyme(s) :
Synthware F352440M, Filter tubes with fritted disc
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.51
Matériaux
colorless tube
glass tube
Joint
joint: ST/NS24/40
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
Synthware F352440M
D.E. du disque
36 mm
Description générale
With two 2 mm glass stopcocks having constant pressure retention devices.
Informations légales
Synthware is a trademark of Kemtech America Inc.