Principaux documents
SYNC161100
Synthware vacuum jacketed distillation column with indents to support packing material
joint: ST/NS 14/20, jacket H 130 mm
Synonyme(s) :
Synthware 161100, glass distilling column
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.51
Joint
joint: ST/NS14/20
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
Synthware C161100
H de l'enveloppe
130 mm
Description générale
With 12 mm I.D. indentations to support packing material. Vacuum jacketed to improve performance .
Informations légales
Synthware is a trademark of Kemtech America Inc.