Documents
SYNA962440
Synthware distillation drip adapter
joint: ST/NS 24/40, bend: 105 deg
Synonyme(s) :
Synthware A962440, Drip adapter, Distillation adapter
About This Item
Code UNSPSC :
41121815
Nomenclature NACRES :
NB.51
Joint
joint: ST/NS24/40
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
Synthware A962440
Catégories apparentées
Description générale
With an outer joint at top. Tapered drip tube extends into open receivers.
Informations légales
Synthware is a trademark of Kemtech America Inc.