Documents
SYNA352440
Synthware distillation adapter
top joint: ST/NS 24/40, bottom joint: ST/NS 24/40, side joint: 24/40, side arm angle: 75 deg
Synonyme(s) :
Synthware A352440, adapter, sidearm adapter
About This Item
Code UNSPSC :
41121815
Nomenclature NACRES :
NB.51
Joint
bottom joint: ST/NS24/40
side joint: 24/40
joint: ST/NS top24/40
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
Synthware A352440
Catégories apparentées
Description générale
For use as a component in atmospheric or vacuum distillations.
Informations légales
Synthware is a trademark of Kemtech America Inc.