Principaux documents
SYNA342942
Synthware twin hose connecting adapter
joint: ST/NS 29/42
Synonyme(s) :
Synthware A342942, Adapter, twin connecting hose
About This Item
Code UNSPSC :
41121815
Nomenclature NACRES :
NB.51
Joint
joint: ST/NS29/42
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
Synthware A342942
Catégories apparentées
Description générale
Adapter has two hose connections 180° from one another to permit a flow of inert gas over the apparatus.
Informations légales
Synthware is a trademark of Kemtech America Inc.