Principaux documents
PL800GB-7
Plas-Labs butyl glove box gloves
for 1 piece glove set, size 7 (for 8 in. ports)
About This Item
Code UNSPSC :
46181504
Nomenclature NACRES :
NB.35
Matériaux
butyl rubber
Taille
7 (for 8 in. ports)
Caractéristiques
reusable
Conditionnement
pack of 1 pair
Fabricant/nom de marque
Plas-Labs 800GB-7
Description générale
Plas-Labs black butyl rubber gloves for highest impermeability to water vapor, gases, and toxic fluids
Note: Order sleeves, gloves, and glove over sleeve connectors separately.