Principaux documents
DWK11-6200-S
Wheaton® Microliter Vial
100 EACH
104,00 $
104,00 $
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Changer de vue
100 EACH
104,00 $
About This Item
Code UNSPSC :
41122001
Nomenclature NACRES :
NB.11
104,00 $
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Matériaux
V-bottom polypropylene bottom
amber borosilicate glass Type 1 vial (Silanized, with Marking Patch)
Conditionnement
pkg of 100 ea
Fabricant/nom de marque
WHEATON® 11-6200-S
P × H
12 mm × 32 mm
Taille du bouchon
11 mm
Volume
2.0 mL
Application
Chromatography
Caractéristiques et avantages
  • Double ring finish for added closure performance.
  • Approved for LC & GC.
  • Where noted, Certified Class 10,000 clean room packed enhances data integrity.
Informations légales
WHEATON is a registered trademark of Wheaton Industries, Inc.
Contenu apparenté
Explore our protocol for suspension primary hepatocyte cell culture. Discover our primary human hepatocytes and plating and maintenance media formulations.
Questions
  1. Does this product come with lids?
    1 answer
    1. This product comes with a 11 mm Snap Cap.